accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80-39-7

IUPAC Name: N-ethyl-4-methylbenzene-1-sulfonamide
Synonyms: N-ethyl-4-methylbenzenesulfonamide
Molecular Weight (g/mol): 199.27
Molecular Formula: C9H13NO2S
InChi Key: OHPZPBNDOVQJMH-UHFFFAOYSA-N
SMILES: CCNS(=O)(=O)C1=CC=C(C)C=C1
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)