accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COHO(E)

CAS RN 80-59-1

IUPAC Name: (2E)-2-methylbut-2-enoic acid
Synonyms: tiglic acid tiglinic acid tiglinsaeure methyl methacrylic acid methylbutenoicacid cevadic acid tiglinate α,β-dimethyl acrylic acid epsilon-tiglic acid epsilon-tiglate
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: UIERETOOQGIECD-ONEGZZNKSA-N
SMILES: C\C=C(/C)C(O)=O
Percent Purity 
  • (3)
  • (3)
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)