CAS RN 80-69-3
IUPAC Name:
2-hydroxypropanedioic acid
Synonyms:
tartronic acid
Molecular Weight (g/mol):
120.06
Molecular Formula:
C3H4O5
InChi Key:
ROBFUDYVXSDBQM-UHFFFAOYSA-N
SMILES:
OC(C(O)=O)C(O)=O
1
–
1
of
1
results
1
Promotions Available
Tartronic acid, 97%
Tartronic acid, 97%
CAS: 80-69-3 Molecular Formula: C3H4O5 Molecular Weight (g/mol): 120.06 MDL Number: MFCD00004237 InChI Key: ROBFUDYVXSDBQM-UHFFFAOYSA-N Synonym: tartronic acid,hydroxymalonic acid,2-hydroxymalonic acid,propanedioic acid, hydroxy,tartronicacid,2-tartronic acid,hydroxypropanedioic acid,2-hydroxymalonate,malonic acid, hydroxy,hydroxymalonate PubChem CID: 45 ChEBI: CHEBI:16513 IUPAC Name: 2-hydroxypropanedioic acid SMILES: C(C(=O)O)(C(=O)O)O