accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3OHH3CH3C

CAS RN 80-97-7

IUPAC Name: 9a,11a-dimethyl-1-(6-methylheptan-2-yl)-hexadecahydro-1H-cyclopenta[a]phenanthren-7-ol
Synonyms: coprostanol
Molecular Weight (g/mol): 388.68
Molecular Formula: C27H48O
InChi Key: QYIXCDOBOSTCEI-UHFFFAOYNA-N
SMILES: CC(C)CCCC(C)C1CCC2C3CCC4CC(O)CCC4(C)C3CCC12C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results