CAS RN 8000-34-8
IUPAC Name:
methyl 2-amino-3-(1H-imidazol-5-yl)propanoate hydrochloride
Synonyms:
clove oil
Molecular Weight (g/mol):
205.64
Molecular Formula:
C7H12ClN3O2
InChi Key:
VEEIFXWJNCAVEQ-UHFFFAOYNA-N
SMILES:
Cl.COC(=O)C(N)CC1=CN=CN1
1
–
1
of
1
results
1
Clove Oil, FCC, 85%, Spectrum™ Chemical
CAS: 8000-34-8 Molecular Formula: C7H12ClN3O2 Molecular Weight (g/mol): 205.64 InChI Key: VEEIFXWJNCAVEQ-UHFFFAOYNA-N IUPAC Name: methyl 2-amino-3-(1H-imidazol-5-yl)propanoate hydrochloride SMILES: Cl.COC(=O)C(N)CC1=CN=CN1