CAS RN 8002-43-5
Thermo Scientific Chemicals L-alpha-Lecithin, granular, from soybean oil
CAS: 8002-43-5 Molecular Formula: C42H80NO8P Molecular Weight (g/mol): 750 MDL Number: MFCD00082428 InChI Key: JLPULHDHAOZNQI-JLOPVYAASA-N Synonym: 3-sn-phosphatidylcholine,1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine,1,2-diacyl-sn-glycero-3-phosphocholine,3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r,l-,a-lecithin,2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine,a-phosphatidylcholine solution,l-alpha-phosphatidylcholine solution,2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine PubChem CID: 16213884 ChEBI: CHEBI:86658 IUPAC Name: [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Thermo Scientific Chemicals Lecithin, Refined
CAS: 8002-43-5 Molecular Formula: C42H80NO8P Molecular Weight (g/mol): 758.075 MDL Number: MFCD00147406 InChI Key: JLPULHDHAOZNQI-JLOPVYAASA-N Synonym: 3-sn-phosphatidylcholine,1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine,1,2-diacyl-sn-glycero-3-phosphocholine,3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r,l-,a-lecithin,2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine,a-phosphatidylcholine solution,l-alpha-phosphatidylcholine solution,2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine PubChem CID: 16213884 ChEBI: CHEBI:86658 IUPAC Name: [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Lecithin from Soybean, TCI America™
CAS: 8002-43-5 Molecular Formula: C42H80NO8P Molecular Weight (g/mol): 758.075 MDL Number: MFCD00147406 InChI Key: JLPULHDHAOZNQI-JLOPVYAASA-N Synonym: 3-sn-phosphatidylcholine,1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine,1,2-diacyl-sn-glycero-3-phosphocholine,3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r,l-,a-lecithin,2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine,a-phosphatidylcholine solution,l-alpha-phosphatidylcholine solution,2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine PubChem CID: 16213884 ChEBI: CHEBI:86658 IUPAC Name: [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Lecithin (Vegetable/Laboratory), Fisher Chemical
CAS: 8002-43-5 Molecular Formula: C42H80NO8P Molecular Weight (g/mol): 758.075 MDL Number: MFCD00131449 InChI Key: JLPULHDHAOZNQI-JLOPVYAASA-N Synonym: 3-sn-phosphatidylcholine,1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine,1,2-diacyl-sn-glycero-3-phosphocholine,3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r,l-,a-lecithin,2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine,a-phosphatidylcholine solution,l-alpha-phosphatidylcholine solution,2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine PubChem CID: 16213884 ChEBI: CHEBI:86658 IUPAC Name: [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Lecithin, 90%, soybean
CAS: 8002-43-5 Molecular Formula: C42H80NO8P Molecular Weight (g/mol): 758.075 MDL Number: MFCD00147406 InChI Key: JLPULHDHAOZNQI-JLOPVYAASA-N Synonym: 3-sn-phosphatidylcholine,1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine,1,2-diacyl-sn-glycero-3-phosphocholine,3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r,l-,a-lecithin,2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine,a-phosphatidylcholine solution,l-alpha-phosphatidylcholine solution,2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine PubChem CID: 16213884 ChEBI: CHEBI:86658 IUPAC Name: [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Lecithin, ∽60%, MP Biomedicals™
CAS: 8002-43-5 Molecular Formula: C42H80NO8P Molecular Weight (g/mol): 758.075 InChI Key: JLPULHDHAOZNQI-JLOPVYAASA-N Synonym: 3-sn-phosphatidylcholine,1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine,1,2-diacyl-sn-glycero-3-phosphocholine,3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r,l-,a-lecithin,2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine,a-phosphatidylcholine solution,l-alpha-phosphatidylcholine solution,2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine PubChem CID: 16213884 ChEBI: CHEBI:86658 IUPAC Name: [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Lecithin, MP Biomedicals™
CAS: 8002-43-5 Molecular Formula: C42H80NO8P Molecular Weight (g/mol): 758.075 InChI Key: JLPULHDHAOZNQI-JLOPVYAASA-N Synonym: 3-sn-phosphatidylcholine,1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine,1,2-diacyl-sn-glycero-3-phosphocholine,3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r,l-,a-lecithin,2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine,a-phosphatidylcholine solution,l-alpha-phosphatidylcholine solution,2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine PubChem CID: 16213884 ChEBI: CHEBI:86658 IUPAC Name: [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Lecithin from Egg, MP Biomedicals™
Synonym: 1,2-Diacyl-sn-glycero-3-phosphocholine,3-sn-Phosphatidylcholine,L-α-Lecithin,Azolectin
Lecithin, Granular, Food Grade, Spectrum™ Chemical
CAS: 8002-43-5 Molecular Formula: C42H80NO8P Molecular Weight (g/mol): 758.08 InChI Key: JLPULHDHAOZNQI-UHFFFAOYNA-N IUPAC Name: (2-{[3-(hexadecanoyloxy)-2-(octadeca-9,12-dienoyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Lecithin, 99.2%, MP Biomedicals™
CAS: 8002-43-5 Molecular Formula: C42H80NO8P Molecular Weight (g/mol): 758.075 InChI Key: JLPULHDHAOZNQI-JLOPVYAASA-N Synonym: 3-sn-phosphatidylcholine,1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine,1,2-diacyl-sn-glycero-3-phosphocholine,3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r,l-,a-lecithin,2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine,a-phosphatidylcholine solution,l-alpha-phosphatidylcholine solution,2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine PubChem CID: 16213884 ChEBI: CHEBI:86658 IUPAC Name: [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Lecithin, 60%, egg, Thermo Scientific Chemicals
CAS: 8002-43-5 Molecular Formula: C42H80NO8P Molecular Weight (g/mol): 758.075 MDL Number: MFCD00131449 InChI Key: JLPULHDHAOZNQI-JLOPVYAASA-N Synonym: 3-sn-phosphatidylcholine,1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine,1,2-diacyl-sn-glycero-3-phosphocholine,3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r,l-,a-lecithin,2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine,a-phosphatidylcholine solution,l-alpha-phosphatidylcholine solution,2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine PubChem CID: 16213884 ChEBI: CHEBI:86658 IUPAC Name: [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Lecithin, MP Biomedicals™
CAS: 8002-43-5 Molecular Formula: C42H80NO8P Molecular Weight (g/mol): 758.075 InChI Key: JLPULHDHAOZNQI-JLOPVYAASA-N Synonym: 3-sn-phosphatidylcholine,1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine,1,2-diacyl-sn-glycero-3-phosphocholine,3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r,l-,a-lecithin,2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine,a-phosphatidylcholine solution,l-alpha-phosphatidylcholine solution,2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine PubChem CID: 16213884 ChEBI: CHEBI:86658 IUPAC Name: [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate SMILES: CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC
Medisca Lecithin (Granular)
Lecithin comes as golden to light tan granules with a neutral odor. It is hygroscopic.
L-α-Egg-phosphatidylcholine, 25 wt.% solution in chloroform, Thermo Scientific™
CAS: 8002-43-5 Molecular Formula: CHCl3 Molecular Weight (g/mol): 119.37 MDL Number: MFCD00000826 InChI Key: HEDRZPFGACZZDS-UHFFFAOYSA-N Synonym: 3-sn-phosphatidylcholine,1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine,1,2-diacyl-sn-glycero-3-phosphocholine,3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r,l-,a-lecithin,2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine,a-phosphatidylcholine solution,l-alpha-phosphatidylcholine solution,2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine PubChem CID: 131706484 IUPAC Name: trichloromethane SMILES: ClC(Cl)Cl