accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOH3CCH3H3CCH3H2CCH3OOOH3CCH3CH3OOH3CH2C(E/Z)(E/Z)(E/Z)

CAS RN 8003-34-7

IUPAC Name: 2-methyl-4-oxo-3-(penta-2,4-dien-1-yl)cyclopent-2-en-1-yl 2,2-dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropane-1-carboxylate; 2-methyl-4-oxo-3-(penta-2,4-dien-1-yl)cyclopent-2-en-1-yl 3-(3-methoxy-2-methyl-3-oxoprop-1-en-1-yl)-2,2-dimethylcyclopropane-1-carboxylate
Synonyms: pyrethrins
Molecular Weight (g/mol): 700.91
Molecular Formula: C43H56O8
InChi Key: VXSIXFKKSNGRRO-UHFFFAOYNA-N
SMILES: CC(C)=CC1C(C(=O)OC2CC(=O)C(CC=CC=C)=C2C)C1(C)C.COC(=O)C(C)=CC1C(C(=O)OC2CC(=O)C(CC=CC=C)=C2C)C1(C)C
Percent Purity 
  • (1)
Quantity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results