accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOSOONSOOOOO

CAS RN 8004-92-0

IUPAC Name: disodium 2-(1,3-dioxo-2,3-dihydro-1H-inden-2-yl)quinoline-6,8-disulfonate
Synonyms: quinoline yellow WS
Molecular Weight (g/mol): 477.37
Molecular Formula: C18H9NNa2O8S2
InChi Key: FZUOVNMHEAPVBW-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC(=C2N=C(C=CC2=C1)C1C(=O)C2=CC=CC=C2C1=O)S([O-])(=O)=O
Grade 
  • (2)
  • (4)
  • (1)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (4)
  • (2)
Color 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (4)
  • (2)
  • (5)