CAS RN 8006-64-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Promotions Available
Turpentine, Rectified, Spectrum™ Chemical
Turpentine, Rectified, Spectrum™ Chemical
2
Sigma Aldrich Oil of turpentine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.