accessibility menu, dialog, popup

UserName

Viewing profile as user:
A(a)

CAS RN 8006-90-4

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: —
SMILES: —
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
Grade 
  • (4)
  • (5)
Physical Form 
  • (8)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (9)
  • (1)
  • (1)
  • (9)
  • (8)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (4)
  • (5)
  • (8)
  • (1)