accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 8007-43-0

IUPAC Name: bis((2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol); tris((9Z)-octadec-9-enoic acid)
Synonyms: sorbitan sesquioleate 倍半油酸山梨坦
Molecular Weight (g/mol): 1211.75
Molecular Formula: C66H130O18
InChi Key: CUNWUEBNSZSNRX-RKGWDQTMSA-N
SMILES: OC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.OC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO.CCCCCCCC\C=C/CCCCCCCC(O)=O.CCCCCCCC\C=C/CCCCCCCC(O)=O.CCCCCCCC\C=C/CCCCCCCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results