CAS RN 80102-25-6
CAS RN 80102-25-6
IUPAC Name:
2-{[(tert-butoxy)carbonyl]amino}-3-(3-chlorophenyl)propanoic acid
Synonyms:
2-[(tert-butoxycarbonyl)amino]-3-(3-chlorophenyl)propanoic acid
Molecular Weight (g/mol):
299.75
Molecular Formula:
C14H18ClNO4
InChi Key:
RCZHBTHQISEPPP-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(CC1=CC(Cl)=CC=C1)C(O)=O
1
–
1
of
1
results
1
Advanced Chem Tech Boc-D-Phe 3-Cl -OH, Advanced ChemTech
CAS: 80102-25-6 Molecular Formula: C14H18ClNO4 Molecular Weight (g/mol): 299.75 InChI Key: RCZHBTHQISEPPP-UHFFFAOYNA-N IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-(3-chlorophenyl)propanoic acid SMILES: CC(C)(C)OC(=O)NC(CC1=CC(Cl)=CC=C1)C(O)=O