accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80126-52-9

IUPAC Name: 2-amino-3-(3-chlorophenyl)propanoic acid
Synonyms: 2-amino-3-(3-chlorophenyl)propanoic acid
Molecular Weight (g/mol): 199.63
Molecular Formula: C9H10ClNO2
InChi Key: JJDJLFDGCUYZMN-UHFFFAOYNA-N
SMILES: NC(CC1=CC(Cl)=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)