accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(S)O(R)OO(S)OH(R)OH(R)OH

CAS RN 80154-34-3

IUPAC Name: 2-{[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}benzaldehyde
Synonyms: 2-{[(2R,3S,4R,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}benzaldehyde
Molecular Weight (g/mol): 284.26
Molecular Formula: C13H16O7
InChi Key: BGOFCVIGEYGEOF-WJTVCTBASA-N
SMILES: OC[C@@H]1O[C@H](OC2=CC=CC=C2C=O)[C@@H](O)[C@H](O)[C@H]1O
Percent Purity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)