accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOPOOOHHn

CAS RN 8017-16-1

IUPAC Name: Polyphosphoric acid
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (HO3P)n
InChi Key: NBIIXXVUZAFLBC-UHFFFAOYSA-N
SMILES: [H]OP(O)(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)