accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 8021-39-4

IUPAC Name: 2-methoxyphenol
Synonyms: guaiacol methylcatechol methyl catechol guiacol guaicol guajacol catechol, methyl o-guaiacol guajakol guajol
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: LHGVFZTZFXWLCP-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results