accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80210-62-4

IUPAC Name: (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Synonyms: cefpodoxime cefpodoxime (substance) cpdx-PR cefpodoxime sodium cefpodoxime (product) cefpodoxime preparation cefpodoxime proxetil otreon cefpodoxime sodium salt cefpodoximum
Molecular Weight (g/mol): 427.45
Molecular Formula: C15H17N5O6S2
InChi Key: WYUSVOMTXWRGEK-HBWVYFAYSA-N
SMILES: COCC1=C(N2[C@H](SC1)[C@H](NC(=O)C(=N/OC)\C1=CSC(N)=N1)C2=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results