accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80370-42-9

IUPAC Name: ethyl 2-formyl-3-oxopropanoate
Synonyms: ethyl 2-formyl-3-oxopropanoate
Molecular Weight (g/mol): 144.13
Molecular Formula: C6H8O4
InChi Key: HMFLBGNCDZYITR-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C=O)C=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results