accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 803727-09-5

IUPAC Name: 4-(5-phenylthiophen-2-yl)-N,N-bis[4-(5-phenylthiophen-2-yl)phenyl]aniline
Synonyms: 4-(5-phenylthiophen-2-yl)-N,N-bis[4-(5-phenylthiophen-2-yl)phenyl]aniline
Molecular Weight (g/mol): 719.98
Molecular Formula: C48H33NS3
InChi Key: UJZRJDHTDMTGAP-UHFFFAOYSA-N
SMILES: S1C(=CC=C1C1=CC=C(C=C1)N(C1=CC=C(C=C1)C1=CC=C(S1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(S1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results