accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80384-27-6

IUPAC Name: (2R,3S)-2-{[(benzyloxy)carbonyl]amino}-3-hydroxybutanoic acid
Synonyms: (2R,3S)-2-{[(benzyloxy)carbonyl]amino}-3-hydroxybutanoic acid|Z-D-THR-OH
Molecular Weight (g/mol): 253.25
Molecular Formula: C12H15NO5
InChi Key: IPJUIRDNBFZGQN-WCBMZHEXSA-N
SMILES: C[C@H](O)[C@@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results