CAS RN 804560-00-7
IUPAC Name:
2-methyl-9,10-bis(naphthalen-2-yl)anthracene
Synonyms:
2-methyl-9,10-bis(naphthalen-2-yl)anthracene
Molecular Weight (g/mol):
444.58
Molecular Formula:
C35H24
InChi Key:
HNWFFTUWRIGBNM-UHFFFAOYSA-N
SMILES:
CC1=CC=C2C(=C1)C(C1=CC=C3C=CC=CC3=C1)=C1C=CC=CC1=C2C1=CC=C2C=CC=CC2=C1
1
–
1
of
1
results
1
2-Methyl-9,10-di(2-naphthyl)anthracene 97.0+%, TCI America™
CAS: 804560-00-7 Molecular Formula: C35H24 Molecular Weight (g/mol): 444.58 MDL Number: MFCD12022447 InChI Key: HNWFFTUWRIGBNM-UHFFFAOYSA-N PubChem CID: 53403806 IUPAC Name: 2-methyl-9,10-bis(naphthalen-2-yl)anthracene SMILES: CC1=CC=C2C(=C1)C(C1=CC=C3C=CC=CC3=C1)=C1C=CC=CC1=C2C1=CC=C2C=CC=CC2=C1