accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2BOHHO

CAS RN 80460-73-7

IUPAC Name: (4-aminophenyl)boronic acid
Synonyms: 4-aminophenylboronic acid
Molecular Weight (g/mol): 136.95
Molecular Formula: C6H8BNO2
InChi Key: MKPDAJWEBQRQCO-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)B(O)O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)