accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80466-80-4

IUPAC Name: dimethyl-1,3-thiazole-5-sulfonyl chloride
Synonyms: dimethyl-1,3-thiazole-5-sulfonyl chloride
Molecular Weight (g/mol): 211.68
Molecular Formula: C5H6ClNO2S2
InChi Key: GFFJSTHQILQFNQ-UHFFFAOYSA-N
SMILES: CC1=NC(C)=C(S1)S(Cl)(=O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)