accessibility menu, dialog, popup

UserName

Viewing profile as user:
CdCdCdCdCdCdCdCdCdCdCdCdSnSn

CAS RN 8049-22-7

IUPAC Name: dodecacadmium ditin
Synonyms: dodecacadmium ditin
Molecular Weight (g/mol): 1586.39
Molecular Formula: Cd12Sn2
InChi Key: OIBLPQFTAJDRMC-UHFFFAOYSA-N
SMILES: [Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Cd].[Sn].[Sn]
Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results