accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNNNN

CAS RN 80510-03-8

IUPAC Name: 1-[bis(1H-pyrazol-1-yl)methyl]-1H-pyrazole
Synonyms: 1-[bis(pyrazol-1-yl)methyl]pyrazole
Molecular Weight (g/mol): 214.23
Molecular Formula: C10H10N6
InChi Key: QOHVIMQQEFHOLE-UHFFFAOYSA-N
SMILES: C1=CN(N=C1)C(N1C=CC=N1)N1C=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results