accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80514-65-4

IUPAC Name: 2-[(triphenylmethyl)amino]propanoic acid; diethylamine
Synonyms: 2-[(triphenylmethyl)amino]propanoic acid; diethylamine
Molecular Weight (g/mol): 404.55
Molecular Formula: C26H32N2O2
InChi Key: PWGPLCPGMCXDCL-UHFFFAOYNA-N
SMILES: CCNCC.CC(NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)