accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 8052-41-3

IUPAC Name: 6-bromohexyl acetate
Synonyms: 0.1% w/w benzene
Molecular Weight (g/mol): 223.11
Molecular Formula: C8H15BrO2
InChi Key: IMSPZLCTPSXORJ-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCCCBr
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)