accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80531-13-1

IUPAC Name: 1-benzyl-1H-indole-5-carbonitrile
Synonyms: 1-benzylindole-5-carbonitrile
Molecular Weight (g/mol): 232.29
Molecular Formula: C16H12N2
InChi Key: XZCSETGZSGBGCI-UHFFFAOYSA-N
SMILES: N#CC1=CC=C2N(CC3=CC=CC=C3)C=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results