accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80548-31-8

IUPAC Name: (2-hydroxyethyl)[(1R)-1-phenylethyl]azanium
Synonyms: (2-hydroxyethyl)[(1R)-1-phenylethyl]azanium
Molecular Weight (g/mol): 166.24
Molecular Formula: C10H16NO
InChi Key: GXIWMXAAPLZOBY-SECBINFHSA-O
SMILES: C[C@@H]([NH2+]CCO)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results