accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHOHOH3CH3CCH3H3COH(E/Z)(E/Z)

CAS RN 80557-12-6

IUPAC Name: 2,4-dihydroxy-6-methyl-3-(3,7,11-trimethyldodeca-2,6,10-trien-1-yl)benzoic acid
Synonyms: 2,4-dihydroxy-6-methyl-3-(3,7,11-trimethyldodeca-2,6,10-trien-1-yl)benzoic acid
Molecular Weight (g/mol): 372.51
Molecular Formula: C23H32O4
InChi Key: QPIZDZGIXDKCRC-UHFFFAOYSA-N
SMILES: CC(C)=CCCC(C)=CCCC(C)=CCC1=C(O)C=C(C)C(C(O)=O)=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results