accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 8065-48-3

CH3OPOOH3CSSH3CCH3OPSOH3COSH3C

CAS RN 8065-48-3

IUPAC Name: O,O-diethyl O-2-(ethylsulfanyl)ethyl phosphorothioate; diethyl {[2-(ethylsulfanyl)ethyl]sulfanyl}phosphonate
Synonyms: systox
Molecular Weight (g/mol): 516.66
Molecular Formula: C16H38O6P2S4
InChi Key: FAXIJTUDSBIMHY-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)SCCSCC.CCOP(=S)(OCC)OCCSCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results