accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3N

CAS RN 80673-00-3

IUPAC Name: 3-[2-(trimethylsilyl)ethynyl]pyridine
Synonyms: 3-[2-(trimethylsilyl)ethynyl]pyridine
Molecular Weight (g/mol): 175.31
Molecular Formula: C10H13NSi
InChi Key: MTMBLPOPETZZRX-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC1=CN=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)