accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80745-09-1

CH3SOOHNNOHOHNOONH2H3CH3COH3CNH2(E/Z)

CAS RN 80745-09-1

IUPAC Name: 5-{[amino(4-methoxy-2,3,6-trimethylbenzenesulfonamido)methylidene]amino}-2-{[(benzyloxy)carbonyl]amino}pentanoic acid; cyclohexanamine
Synonyms: 5-{[amino(4-methoxy-2,3,6-trimethylbenzenesulfonamido)methylidene]amino}-2-{[(benzyloxy)carbonyl]amino}pentanoic acid; cyclohexylamine
Molecular Weight (g/mol): 619.78
Molecular Formula: C30H45N5O7S
InChi Key: MDRGEGBEEQDQPG-UHFFFAOYNA-N
SMILES: NC1CCCCC1.COC1=C(C)C(C)=C(C(C)=C1)S(=O)(=O)NC(N)=NCCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)