accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 80789-75-9

IUPAC Name: 1-(4-methylphenyl)cyclopentane-1-carboxylic acid
Synonyms: 1-(4-methylphenyl)cyclopentane-1-carboxylic acid
Molecular Weight (g/mol): 204.27
Molecular Formula: C13H16O2
InChi Key: YKDWTRWSHHGVII-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1(CCCC1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results