accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 808-57-1

IUPAC Name: 2,3,6,7,10,11-hexamethoxytriphenylene
Synonyms: 2,3,6,7,10,11-hexamethoxytriphenylene
Molecular Weight (g/mol): 408.45
Molecular Formula: C24H24O6
InChi Key: TXROZCSFVVIBFI-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C(=C1)C1=CC(OC)=C(OC)C=C1C1=CC(OC)=C(OC)C=C21
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results