accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80866-75-7

IUPAC Name: (3-methyl-4-nitrophenyl)methanol
Synonyms: (3-methyl-4-nitrophenyl)methanol
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: KOVQGYQQVNCUBR-UHFFFAOYSA-N
SMILES: CC1=CC(CO)=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results