accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80866-87-1

IUPAC Name: 3,3,3-trifluoro-2-(2-methoxyphenyl)propanenitrile
Synonyms: 3,3,3-trifluoro-2-(2-methoxyphenyl)propanenitrile
Molecular Weight (g/mol): 215.18
Molecular Formula: C10H8F3NO
InChi Key: SWVVGQSOWXDMQT-UHFFFAOYNA-N
SMILES: COC1=CC=CC=C1C(C#N)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results