accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNBr

CAS RN 808744-34-5

IUPAC Name: 7-bromoimidazo[1,2-a]pyridine
Synonyms: 7-bromoimidazo[1,2-a]pyridine
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: OASOJRLJBDCVNU-UHFFFAOYSA-N
SMILES: BrC1=CC2=NC=CN2C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)