accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(S)(S)OH(R)CH3(S)OCH3(R)OO(R)H3C(R)HOH3COH3C(S)CH3(R)HO(S)CH3(R)HO(R)O(R)(R)OH(S)ONH2O(R)H3CO(S)CH3(R)OH3CH3C(E)(E)(Z)(E)(E)

CAS RN 80890-47-7

IUPAC Name: (2R,3S,4R,6R)-6-{[(2R,4R,5S,6R)-2-[(2S,3R,4S)-4-[(2R,3S,4E,6E,9R,10S,11S,12R,13R,14E,16Z)-11-ethyl-10,12-dihydroxy-3,17-dimethoxy-7,9,13,15-tetramethyl-18-oxo-1-oxacyclooctadeca-4,6,14,16-tetraen-2-yl]-3-hydroxypentan-2-yl]-2-hydroxy-5-methyl-6-[(1E)-prop-1-en-1-yl]oxan-4-yl]oxy}-4-hydroxy-2-methyloxan-3-yl carbamate
Synonyms: (2R,3S,4R,6R)-6-{[(2R,4R,5S,6R)-2-[(2S,3R,4S)-4-[(2R,3S,4E,6E,9R,10S,11S,12R,13R,14E,16Z)-11-ethyl-10,12-dihydroxy-3,17-dimethoxy-7,9,13,15-tetramethyl-18-oxo-1-oxacyclooctadeca-4,6,14,16-tetraen-2-yl]-3-hydroxypentan-2-yl]-2-hydroxy-5-methyl-6-[(1E)-prop-1-en-1-yl]oxan-4-yl]oxy}-4-hydroxy-2-methyloxan-3-yl carbamate
Molecular Weight (g/mol): 866.1
Molecular Formula: C46H75NO14
InChi Key: DJZCTUVALDDONK-HQMSUKCRSA-N
SMILES: CC[C@H]1[C@@H](O)[C@H](C)C\C(C)=C\C=C\[C@H](OC)[C@H](OC(=O)\C(OC)=C\C(\C)=C\[C@@H](C)[C@H]1O)[C@@H](C)[C@@H](O)[C@H](C)[C@@]1(O)C[C@@H](O[C@H]2C[C@@H](O)[C@H](OC(N)=O)[C@@H](C)O2)[C@H](C)[C@H](O1)\C=C\C
Quantity 
  • (1)
Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results