accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 80938-67-6

IUPAC Name: 4-methyl-[1,1'-biphenyl]-3-amine
Synonyms: 4-methyl-[1,1'-biphenyl]-3-amine
Molecular Weight (g/mol): 183.25
Molecular Formula: C13H13N
InChi Key: YLKSTPDTTKOSIL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1N)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results