CAS RN 81-42-5
IUPAC Name:
1,4-diamino-2,3-dichloro-9,10-dihydroanthracene-9,10-dione
Synonyms:
1,4-diamino-2,3-dichloroanthracene-9,10-dione
Molecular Weight (g/mol):
307.13
Molecular Formula:
C14H8Cl2N2O2
InChi Key:
KZYAYVSWIPZDKL-UHFFFAOYSA-N
SMILES:
NC1=C(Cl)C(Cl)=C(N)C2=C1C(=O)C1=CC=CC=C1C2=O
1
–
1
of
1
results
1
1,4-Diamino-2,3-dichloroanthraquinone 93.0+%, TCI America™
CAS: 81-42-5 Molecular Formula: C14H8Cl2N2O2 Molecular Weight (g/mol): 307.13 MDL Number: MFCD00035693 InChI Key: KZYAYVSWIPZDKL-UHFFFAOYSA-N Synonym: Disperse Violet 28 PubChem CID: 65731 IUPAC Name: 1,4-diamino-2,3-dichloro-9,10-dihydroanthracene-9,10-dione SMILES: NC1=C(Cl)C(Cl)=C(N)C2=C1C(=O)C1=CC=CC=C1C2=O