accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOHOO

CAS RN 81-48-1

IUPAC Name: 1-hydroxy-4-[(4-methylphenyl)amino]-9,10-dihydroanthracene-9,10-dione
Synonyms: solvent violet 13 alizurol purple
Molecular Weight (g/mol): 329.36
Molecular Formula: C21H15NO3
InChi Key: LJFWQNJLLOFIJK-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC2=CC=C(O)C3=C2C(=O)C2=CC=CC=C2C3=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)