accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 81-93-6

IUPAC Name: 3,7-diamino-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Synonyms: phenosafranine phenosafranine, chloride phenosafranin
Molecular Weight (g/mol): 322.8
Molecular Formula: C18H15ClN4
InChi Key: SOUHUMACVWVDME-UHFFFAOYSA-N
SMILES: [Cl-].NC1=CC2=[N+](C3=CC=CC=C3)C3=CC(N)=CC=C3N=C2C=C1
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)