accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 81-96-9

IUPAC Name: 14-bromotetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1(17),2,4,6,9,11,13,15-octaen-8-one
Synonyms: 3-bromobenzanthrone
Molecular Weight (g/mol): 309.16
Molecular Formula: C17H9BrO
InChi Key: WVECFEIAZAKUNF-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=C3C(=O)C4=CC=CC=C4C(C=C1)=C23
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)