accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 81028-96-8

IUPAC Name: 2-{[(7-hydroxy-4-methyl-2-oxo-2H-chromen-6-yl)methyl](methyl)amino}acetic acid
Synonyms: {[(7-hydroxy-4-methyl-2-oxochromen-6-yl)methyl](methyl)amino}acetic acid
Molecular Weight (g/mol): 277.28
Molecular Formula: C14H15NO5
InChi Key: KRMRPQPGZHQMDD-UHFFFAOYSA-N
SMILES: CN(CC(O)=O)CC1=C(O)C=C2OC(=O)C=C(C)C2=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results