accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 81096-93-7

IUPAC Name: 4-(benzyloxy)butane-1,3-diol
Synonyms: 4-(benzyloxy)butane-1,3-diol
Molecular Weight (g/mol): 196.25
Molecular Formula: C11H16O3
InChi Key: ZVVBUAFZGGHYAO-UHFFFAOYNA-N
SMILES: OCCC(O)COCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results