accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 811-68-7

IUPAC Name: trifluoromethanethiol silver
Synonyms: trifluoromethanethiol silver
Molecular Weight (g/mol): 209.94
Molecular Formula: CHAgF3S
InChi Key: SVPQZGWGYCARGG-UHFFFAOYSA-N
SMILES: [Ag].FC(F)(F)S
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results