accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3COOOOHHNCH3CH3

CAS RN 81161-17-3

IUPAC Name: methyl 3-(4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)propanoate hydrochloride
Synonyms: esmolol hydrochloride
Molecular Weight (g/mol): 331.84
Molecular Formula: C16H26ClNO4
InChi Key: GEKNCWBANDDJJL-UHFFFAOYNA-N
SMILES: Cl.COC(=O)CCC1=CC=C(OCC(O)CNC(C)C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results