accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3NCl

CAS RN 81167-60-4

IUPAC Name: 4-tert-butyl-2-chloropyridine
Synonyms: 4-tert-butyl-2-chloropyridine
Molecular Weight (g/mol): 169.65
Molecular Formula: C9H12ClN
InChi Key: UGGNULHQVUWRGQ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(Cl)=NC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results