accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOCH3OCH3OCH3CH3OOOCH3OCH3OCH3Pd(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 811862-77-8

IUPAC Name: bis(1,5-bis(3,5-dimethoxyphenyl)penta-1,4-dien-3-one) palladium
Synonyms: bis(1,5-bis(3,5-dimethoxyphenyl)penta-1,4-dien-3-one) palladium
Molecular Weight (g/mol): 815.22
Molecular Formula: C42H44O10Pd
InChi Key: RTGAJOPJZJDWAX-UHFFFAOYSA-N
SMILES: [Pd].COC1=CC(C=CC(=O)C=CC2=CC(OC)=CC(OC)=C2)=CC(OC)=C1.COC1=CC(C=CC(=O)C=CC2=CC(OC)=CC(OC)=C2)=CC(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results